missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA575268
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
Spécification
| COX11 | |
| Polyclonal | |
| Unconjugated | |
| COX11 | |
| 2010004I09Rik; Cox11; COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; COX11, cytochrome c oxidase copper chaperone; COX11P; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1353, 690300, 69802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P8I6, Q9Y6N1 | |
| COX11 | |
| Synthetic peptide corresponding to amino acids 50-100 of Human COX11. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction