Learn More
CRIP2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-18431JF646
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CRIP2 Polyclonal antibody specifically detects CRIP2 in Human, Mouse samples. It is validated for Western Blot
Spécification
| CRIP2 | |
| Polyclonal | |
| Western Blot | |
| CRIP, CRP-2, CRP2ESP1, cysteine-rich protein 2, Cystein-rich intestinal protein, Protein ESP1 | |
| Recognizes region between residues Between Between 158 and 208.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1397 | |
| Store at 4°C in the dark. | |
| IgG |