Learn More
CRISPR-Cas9 Antibody - BSA Free, Novus Biologicals™
Code produit 18266904 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.025 mg
0.1 mg
Conditionnement:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18266904 0.025 mg 25µg
18268791 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18266904 Fournisseur Novus Biologicals Code fournisseur NBP266379SS
Rabbit Polyclonal Antibody
CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Antigène CRISPR-Cas9
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugué Unconjugated
Dilution Western Blot 2μg/ml
Formule PBS with 0.02% Sodium Azide
Alias de gène Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Espèces hôtes Rabbit
Immunogène Full length S. pyogenes Cas9 protein [UniProt Q99ZW2]
Méthode de purification Affinity Purified
Quantité 0.025 mg
Disciplines de recherche Gene Editing, Gene Knockouts, Gene Therapy
Primaire ou secondaire Primary
Spécificité du test Cas9 protein from Streptococcus pyogene serotype M1.
Espèces cibles Bacteria
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG