Learn More
CRISPR-Cas9 Antibody, Novus Biologicals™
Code produit 18642889 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18642889 100 μg 100µL
18639847 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18642889 Fournisseur Novus Biologicals Code fournisseur NBP305547100ul
Chicken Polyclonal Antibody
CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Antigène CRISPR-Cas9
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000
Formule Supplied as an aliquot of IgY preparation
Alias de gène Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Espèces hôtes Chicken
Immunogène C-terminal region of S. aureus CRISPR-Cas9 expressed in and purified from E. coli.
Méthode de purification IgY purified
Quantité 100 μg
État réglementaire RUO
Disciplines de recherche Gene Editing, Gene Knockouts, Gene Therapy
Primaire ou secondaire Primary
Identification génétique (Entrez) 901176
Espèces cibles Bacteria
Contenu et stockage Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Forme Purified
Isotype IgY