Learn More
CRISPR-Cas9 Antibody (SR1104), Novus Biologicals™
Code produit 18699284 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18699284 50 μg 50µL
18634644 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18699284 Fournisseur Novus Biologicals Code fournisseur NBP32173250ul
Rabbit Monoclonal Antibody
CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in S. aureus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Spécification
Antigène CRISPR-Cas9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1104
Conjugué Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30
Formule PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gène Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from CRISPR-Cas9 (Uniprot #: J7RUA5)
Méthode de purification Affinity purified
Quantité 50 μg
État réglementaire RUO
Disciplines de recherche Gene Editing, Gene Knockouts, Gene Therapy
Primaire ou secondaire Primary
Identification génétique (Entrez) 901176
Espèces cibles S. aureus
Contenu et stockage Store at -20° C. Avoid freeze/thaw cycles.
Forme Purified
Isotype IgG