missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CROCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554346
This item is not returnable.
View return policy
Description
Immunogen sequence: VKRLEKQNLE KDQVNKDLTE KLEALESLRL QEQAALETED GEGLQQTLRD LAQAVLSDSE SGVQLSGSER TADASNGS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
Major structural component of the ciliary rootlet, a cytoskeletal-like structure in ciliated cells which originates from the basal body at the proximal end of a cilium and extends proximally toward the cell nucleus. Furthermore, is required for the correct positioning of the cilium basal body relative to the cell nucleus, to allow for ciliogenesis (PubMed:27623382). Contributes to centrosome cohesion before mitosis (PubMed:16203858). [UniProt].
Specifications
| CROCC | |
| Polyclonal | |
| Unconjugated | |
| CROCC | |
| ciliary rootlet coiled-coil protein; ciliary rootlet coiled-coil, rootletin; CROCC; KIAA0445; ROLT; rootletin; rootletin, ciliary rootlet protein; Tax1-binding protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TZA2 | |
| CROCC | |
| Recombinant protein corresponding to Human CROCC. Recombinant protein control fragment (Product #RP-93571). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction