missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11709-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.Spécification
| CSN1 | |
| Polyclonal | |
| Unconjugated | |
| GPS1 | |
| COPS1, CSN1, G protein pathway suppressor 1, GPS 1, GPS1, Protein MFH, SGN1, Signalosome subunit 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2873 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13098 | |
| GPS1 | |
| CSN1 Fusion Protein Ag2273 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu