missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CT112 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA559717
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: SDGCGADGLR SRVKYGVKTT PESPPYSSGS YDSIKTEVSG CPEDLTVGRA PTADDDDDDH DDHEDNDKMN DSEGMDPERL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
C20orf112, also termed as chromosome 20 open reading frame 112, is a 436 amino acid protein, which consists of a relatively hydrophobic N terminal region and two putative small coiled-coil domains. C20orf112 may likely involve in endocytosis pathway and normal brain development. It also is required to mediate the cell energy response. The function of C20orf112 is still non-known and mechanism is required to further study.
Spécification
| CT112 | |
| Polyclonal | |
| Unconjugated | |
| NOL4L | |
| 4931410H15; 8430427H17Rik; AI429157; C20orf112; C20orf113; dJ1184F4.2; dJ1184F4.4; Gm1636; NOL4L; nucleolar protein 4 like; nucleolar protein 4-like; RGD1563510 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140688 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MY1 | |
| NOL4L | |
| Recombinant protein corresponding to Human CT112. Recombinant protein control fragment (Product #RP-99467). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu