missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CT112 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5113309
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CT112 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
C20orf112, also termed as chromosome 20 open reading frame 112, is a 436 amino acid protein, which consists of a relatively hydrophobic N terminal region and two putative small coiled-coil domains. C20orf112 may likely involve in endocytosis pathway and normal brain development. It also is required to mediate the cell energy response. The function of C20orf112 is still non-known and mechanism is required to further study.
Spécification
| CT112 | |
| Polyclonal | |
| Unconjugated | |
| NOL4L | |
| 4931410H15; 8430427H17Rik; AI429157; C20orf112; C20orf113; dJ1184F4.2; dJ1184F4.4; Gm1636; NOL4L; nucleolar protein 4 like; nucleolar protein 4-like; RGD1563510 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 140688 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96MY1 | |
| NOL4L | |
| Recombinant Human Nucleolar protein 4-like protein (101-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu