missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CL1/Fractalkine Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
512.00€
Spécification
| Antigène | CX3CL1/Fractalkine |
|---|---|
| Concentration | 1.15 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
Description
CX3CL1/Fractalkine Polyclonal specifically detects CX3CL1/Fractalkine in Human, Mouse, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Spécification
| CX3CL1/Fractalkine | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Gerbil | |
| P78423, O35188 | |
| 6376 | |
| A genomic peptide made to an internal region of the human CX3CL protein (within residues 20-150). [Swiss-Prot P78423] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.15 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Chemokine Research, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| CX3CL1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit