missing translation for 'onlineSavingsMsg'
Learn More

CXCL3 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen 500-P105-100UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17894283

  • 239.00€ / 100µg
Expédition estimée: 25-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

AA Sequence of recombinant protein: ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-gamma (CXCL3). Anti-Human GRO-gamma (CXCL3)-specific antibody was purified by affinity chromatography employing an immobilized Human GRO-gamma (CXCL3) matrix. Sandwich ELISA: To detect Human GRO-gamma (CXCL3) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human GRO-gamma (CXCL3) (500-P105Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GRO-gamma (CXCL3). Western Blot: To detect Human GRO-gamma (CXCL3) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human GRO-gamma (CXCL3) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CXCL3 Polyclonal antibody specifically detects CXCL3 in Human samples. It is validated for ELISA, Western Blot
Spécification

Spécification

CXCL3
Polyclonal
PBS with no preservative
P19876
Rabbit
Antigen affinity chromatography
RUO
2921
-20°C
ELISA, Western Blot
Unconjugated
Cxcl3
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3
E.coli-derived Recombinant Human GRO-gamma (CXCL3)
100 μg
Primary
Human
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions