missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-reported
Spécification
Spécification
| Antigène | CXCR1/IL-8RA |
| Applications | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Clone | 42705R |
| Conjugué | Biotin |
| Dilution | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-reported |
| Formule | PBS |
| Alias de gène | CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A |
| Espèces hôtes | Mouse |
| Immunogène | NS0 mouse myeloma cell line transfected with human CXCR1/IL-8 RA, Met1-Leu350, Accession # AAA59159 |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?