missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cy3/Cy5 Recombinant Rabbit Monoclonal Antibody (1D9D3)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA555189
This item is not returnable.
View return policy
Description
Immunogen Sequence: Cy5-NHS-KL H.
Cy3/Cy5 is a widely used gene target for fluorescent labeling in molecular biology research. Cy3 and Cy5 are fluorescent dyes that emit distinct colors when excited by specific wavelengths of light. These dyes are commonly employed in techniques such as fluorescence in situ hybridization (FISH), microarray analysis, and real-time PCR. By attaching Cy3 or Cy5 to gene-specific probes or primers, researchers can visualize and quantify gene expression levels, identify genomic locations, and monitor DNA/RNA interactions. The compatibility of Cy3/Cy5 with various imaging systems and its bright signal make it a valuable tool in many biological applications, aiding in the understanding of gene function and regulation.
Specifications
| Cy3/Cy5 | |
| Recombinant Monoclonal | |
| 1.98 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Chemical compounds corresponding to Cy5. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Peptide Array, ELISA | |
| 1D9D3 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction