missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cy5 Recombinant Rabbit Monoclonal Antibody (0K5H10)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA555190
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cy5 Recombinant Monoclonal Antibody for ARRAY, ELISA
Cy5 is a widely used fluorescent dye in molecular biology research, specifically for gene targeting and labeling applications. It belongs to the cyanine dye family and emits in the red region of the electromagnetic spectrum when excited by an appropriate light source. Cy5 is commonly employed in techniques such as fluorescence in situ hybridization (FISH), microarray analysis, and real-time PCR. By attaching Cy5 to gene-specific probes or primers, researchers can visualize and quantify gene expression levels, identify genomic locations, and study DNA/RNA interactions. Cy5's strong signal, high photostability, and compatibility with various imaging systems make it a valuable tool in molecular biology, facilitating the investigation of gene function and regulation.
Spécification
| Cy5 | |
| Recombinant Monoclonal | |
| 1.4 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.09% sodium azide; pH 7.3 | |
| Chemical compounds corresponding to Cy5. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Peptide Array, ELISA | |
| 0K5H10 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu