Description
CYP4F2 Polyclonal antibody specifically detects CYP4F2 in Human samples. It is validated for Western Blot
Spécification
Spécification
| Antigène | CYP4F2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formule | PBS with 50% glycerol, pH7.3. |
| Alias de gène | CPF2, CYPIVF2, Cytochrome P450 4F2, cytochrome P450, family 4, subfamily F, polypeptide 2, cytochrome P450, subfamily IVF, polypeptide 2, Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega-hydroxylase, Leukotriene-B(4) 20-monooxygenase 1, leukotriene-B(4) omega-hydroxylase 1, leukotriene-B4 20-monooxygenase |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human CYP4F2 (NP_001073.3). AFYDNCRRLRCFPQPPRRNWFWGHQGMVNPTEEGMRVLTQLVATYPQGFKVWMGPISPLLSLCHPDIIRSVINASAAIAPK |
| Méthode de purification | Affinity purified |
| Afficher plus |