missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome c Antibody (7H8.2C12) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-80686
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Drosophila, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready.
Spécification
| Cytochrome c | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 0.05-0.5 ug/ml, Simple Western : 2.5 ug/ml, Flow Cytometry : 1:20-1:2000, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Yamasaki et al (2006)), Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Fujimara et al (1998)), Immunoblotting reported in scientific literature (PMID 16436379), Flow (Cell Surface) reported in scientific literature (PMID 16467206), Flow (Intracellular) reported in scientific literature (Mohr et al (2004)), CyTOF-ready | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Drosophila, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, CyTOF | |
| 7H8.2C12 | |
| Unconjugated | |
| PBS | |
| CYCS | |
| Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991). | |
| 0.1 mg | |
| Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu