Learn More
Cytokeratin 10 Antibody (rKRT10/6923), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-20243-20ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
| Cytokeratin 10 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Recombinant fragment (around aa384-584) of human Cytokeratin 10 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cell Biology, Cellular Markers | |
| 3858 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rKRT10/6923 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |