missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 (KRT10) (Suprabasal Epithelial Marker) Rabbit, Clone: KRT10/1990R, enQuireBio™
Rabbit Monoclonal Antibody
199.00€ - 554.40€
Spécification
| Antigène | Cytokeratin 10 (KRT10) (Suprabasal Epithelial Marker) |
|---|---|
| Clone | KRT10/1990R |
| Concentration | 200μg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
|
15970358
|
EnquireBio
3858-RBM10-P0 |
20 μg | |||||||
|
15980358
|
EnquireBio
3858-RBM10-P1 |
100 μg |
554.40€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Specifications
| Cytokeratin 10 (KRT10) (Suprabasal Epithelial Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Rabbit | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Skin extract of a human Psoriasis patient | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 56.5kDa |
| KRT10/1990R | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 3858 | |
| IgG | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cytoplasmic | |
| A431, HeLa, MCF7 cells. Esophagus or tonsil. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title