missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Mouse anti-Human, Clone: KRT13/2659, Novus Biologicals™
Mouse Monoclonal Antibody
272.00€
Spécification
| Antigène | Cytokeratin 13 |
|---|---|
| Clone | KRT13/2659 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spécification
| Cytokeratin 13 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3860 | |
| IgG1 κ | |
| Protein A or G purified |
| KRT13/2659 | |
| Flow Cytometry 1-2 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| Recombinant full-length human Cytokeratin 13 protein | |
| Primary | |
| Store at 4°C. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit