missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 8/18 Monoclonal Antibody (5D3)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA514088
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA5-14088 targets Cytokeratin 8/18 in immunohistochemistry (paraffin) applications and shows reactivity with Human samples. The MA5-14088 immunogen is cytokeratins purified from breast cancer MCF-7 cells.
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).
Spécification
| Cytokeratin 8/18 | |
| Monoclonal | |
| Conc. Not Determined | |
| Tissue culture supernatant with 0.09% sodium azide | |
| P05783, P05787 | |
| KRT18, KRT8 | |
| Cytokeratins purified from breast cancer MCF-7 cells. | |
| RUO | |
| 3856, 3875 | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 5D3 | |
| Unconjugated | |
| KRT8 | |
| AA960620; AL022697; AU019895; CARD2; cell proliferation-inducing gene 46 protein; CK18; CK-18; CK8; CK-8; CYK18; CYK8; cytokeratin 18; cytokeratin 8; cytokeratin endo A; Cytokeratin endo B; cytokeratin-18; Cytokeratin-8; Endo B; EndoA; K18; K2C8; K8; keratin 18; keratin 18, type I; keratin 8; keratin 8, type II; keratin complex 1, acidic, gene 18; keratin complex 2, basic, gene 8; keratin D; Keratin, type I cytoskeletal 18; keratin, type II cytoskeletal 8; Keratin-18; keratin-8; Kerd; KO; Krt1-18; KRT18; Krt-2.8; Krt2-8; Krt8; PIG46; Type-II keratin Kb8 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu