Learn More
Cytokeratin 8/18 Mouse, Clone: KRT8.18/1346, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio MSM27-1346-P0
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Cytokeratin 8/18 | |
| Cytoplasmic | |
| KRT8.18/1346 | |
| Unconjugated | |
| Mouse | |
| MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma | |
| 20 μg | |
| Primary | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| Cytoskeleton preparation from HeLa cells | |
| Protein A/G Purified | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |