missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytomegalovirus IE 72 kDa Antigen Monoclonal Antibody (1K10)
Mouse Monoclonal Antibody
Marque: Invitrogen™ MA17596
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-7596 detects HCMV from virus samples. The MA1-7596 immunogen is an HCMV IE concentrate. Store short-term at 4°C, long-term at -80°C.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
Spécification
| Cytomegalovirus IE 72 kDa Antigen | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.01% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot, Immunocytochemistry | |
| 1K10 | |
| Unconjugated | |
| CMV | |
| HCMV IE concentrate. | |
| 100 μg | |
| Primary | |
| 4°C short term, -80°C long term | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu