Learn More
D-Dimer Antibody (DD1), Alexa Fluor™ 350, Novus Biologicals™
Code produit 18758030 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.2 mL
Conditionnement:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18758030 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18758030 Fournisseur Novus Biologicals Code fournisseur NB1108374AF350
Mouse Monoclonal Antibody
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human, Monkey, Baboon samples. It is validated for Western Blot, ELISA, Immunoassay
TRUSTED_SUSTAINABILITY
Spécification
Antigène D-Dimer
Applications Western Blot
Classification Monoclonal
Clone DD1
Conjugué Alexa Fluor 350
Formule 50mM Sodium Borate
Alias de gène D Dimer
Espèces hôtes Mouse
Immunogène Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer.
Méthode de purification Protein A purified
Quantité 0.2 mL
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG2a