missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11985-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DCD is a secreted protein that is subsequently processed into mature peptides of distinct biological activities. The C-terminal peptide is constitutively expressed in sweat and has antibacterial and antifungal activities. The N-terminal peptide, also known as diffusible survival evasion peptide, promotes neural cell survival under conditions of severe oxidative stress. A glycosylated form of the N-terminal peptide may be associated with cachexia (muscle wasting) in cancer patients.Spécification
| DCD | |
| Polyclonal | |
| Unconjugated | |
| DCD | |
| AIDD, DCD, DCD 1, dermcidin, DSEP, HCAP, PIF, Preproteolysin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117159 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P81605 | |
| DCD | |
| DCD Fusion Protein Ag2613 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu