missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25203-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
DDHD2, also known as KIAA0725p, is a member of the intracellular phospholipase A1 (PLA1) protein family which comprises a group of enzymes that hydrolyze the sn-1 ester bond of phospholipids, producing 2-acyl-lysophospholipids and fatty acids. DDHD2 prefers phosphatidic acid as substrate and has a role in efficient membrane trafficking from the Golgi apparatus to the plasma membrane. The gene of DDHD2 maps to chromosome 8p11.23, and encodes a 711-amino-acid protein with a calculated molecular mass of 81 kDa. It has been reported that DDHD2 immunoblot analysis of various tissue extracts revealed two bands of 90 and 85 kDa.Spécification
DDHD2 | |
Polyclonal | |
Unconjugated | |
DDHD2 | |
DDHD domain containing 2, DDHD2, KIAA0725, KIAA0725p, Phospholipase DDHD2, SAMWD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23259, 680971, 72108 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94830, Q80Y98 | |
Ddhd2 | |
DDHD2 Fusion Protein Ag18373 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |