missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DEC2/SHARP1 Polyclonal antibody specifically detects DEC2/SHARP1 in Human, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigène | DEC2/SHARP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000 |
| Formule | PBS with 50% glycerol, pH7.3. |
| Alias de gène | basic helix-loop-helix domain containing, class B, 3, basic helix-loop-helix family, member e41, bHLHb3, BHLHB3class E basic helix-loop-helix protein 41, bHLHe41, Class B basic helix-loop-helix protein 3, DEC2Enhancer-of-split and hairy-related protein 1, Differentially expressed in chondrocytes protein 2, SHARP-1, SHARP1hDEC2 |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant fusion protein containing a sequence corresponding to amino acids 120-280 of human BHLHE41 (NP_110389.1). LKSPIQSDLDAFHSGFQTCAKEVLQYLSRFESWTPREPRCVQLINHLHAVATQFLPTPQLLTQQVPLSKGTGAPSAAGSAAAPCLERAGQKLEPLAYCVPVIQRTQPSAELAAENDTDTDSGYGGEAEARPDREKGKGAGASRVTIKQEPPGEDSPAPKRM |
| Méthode de purification | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?