Learn More
PeproTech DEFA1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Marque: PeproTech 500-P126GBT-50UG
Description
AA Sequence of recombinant protein: ACYCRIPACI AGERRYGTCI YQGRLWAFCC. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human NP-1. Anti-Human NP-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human NP-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NP-1 (500-P126G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human NP-1. Western Blot: To detect hNP-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNP-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P126GBT-1MG will be provided as 2 x 500 μg
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 3 by only one amino acid. This gene and the gene encoding defensin, alpha 3 are both subject to copy number variation.
Spécification
| DEFA1 | |
| Polyclonal | |
| Biotin | |
| DEFA1 | |
| alpha-defensin 1; Cryptdin-1; DEF1; Defa1; DEFA1B; DEFA2; Defcr; Defcr1; defensin alpha 1; defensin related cryptdin peptide 1; defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; defensin-1; defensin-related cryptdin peptide; HNP-1; HNP-2; HP 1-56; HP1; HP-1; HP-2; MRS; myeloid-related sequence; neutrophil defensin 1; Neutrophil defensin 2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 1667 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P59665 | |
| DEFA1 | |
| E.coli-derived, 3.4 kDa Recombinant Human NP-1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.