Learn More
Invitrogen™ Desmosome/Hemidesmosome Recombinant Human Monoclonal Antibody (F12)

Human Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA551829
Description
F12 recognizes an unknown polypeptide of desmosomal and hemidesmosomal plaques with a MW of 185 kDa (determined from immunoblotting using bovine tongue epithelium extract as the substrate), and by IEM (immunoelectron microscopy) it was determined that it is the intracellular part of the desmosome that is bound.
Desmosomes and Hemidesmosomes are two types of intercellular junctions that play important roles in maintaining the structural integrity of tissues subjected to mechanical stress or shear forces. Desmosomes connect neighboring cells using cadherins and intermediate filaments, while Hemidesmosomes anchor epithelial cells to the underlying extracellular matrix using integrins. Both junctions are found in tissues like skin, heart, and muscle.
Spécification
| Desmosome/Hemidesmosome | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| F12 was established by fusing peripheral blood lymphocytes derived from a human vulgaris patient with a hetermyeloma cell line. | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunomicroscopy | |
| F12 | |
| Unconjugated | |
| Human | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgM κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.