Learn More
DFF40/CAD Antibody (36A349), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB120-13592MFV500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DFF40/CAD Monoclonal antibody specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot
Spécification
| DFF40/CAD | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 36A349 | |
| Western Blot | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40. | |
| 0.1 mL | |
| Apoptosis | |
| 1677 | |
| Store at 4C in the dark. | |
| IgG |