missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFF40/CAD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Marque: Novus Biologicals NBP1-77009-0.025mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DFF40/CAD Polyclonal specifically detects DFF40/CAD in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| DFF40/CAD | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Rabbit | |
| 40 kDa | |
| 0.025 mg | |
| Apoptosis | |
| 1677 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| O54788 | |
| Dffb | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the center of murine CAD. The immunogen is located within amino acids 190 - 240 of CAD. Amino Acid Squence: YNGSYFDRGAEASSRLCT | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction