missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA558723
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: EAYILFVRKN AIVVLIPKYG LEGTVFFEEK DKPNPQLIYD DEIPSLKIED TVFHVFDKVK VKIMLDSSNL QHQKIRMSLV EPQIPGISIP TDTSN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
Spécification
| DIS3 | |
| Polyclonal | |
| Unconjugated | |
| DIS3 | |
| 2810028N01Rik; bA555G22.1; DIS3; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; DIS3 mitotic control homolog (S. cerevisiae); dis3p; EXOSC11; exosome complex exonuclease RRP44; exosome component 11; KIAA1008; mitotic control protein dis3 homolog; protein DIS3 homolog; RGD1304646; Ribosomal RNA-processing protein 44; RP11-342J4.3; RRP44 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2L1 | |
| DIS3 | |
| Recombinant protein corresponding to Human DIS3. Recombinant protein control fragment (Product #RP-97922). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Spot an opportunity for improvement?Share a Content Correction