Learn More
DLEC/CLEC4C/BDCA-2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-92121-0.1ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DLEC/CLEC4C/BDCA-2 Polyclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Western Blot
Spécification
| DLEC/CLEC4C/BDCA-2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 45-213 of human CLEC4C (NP_569708.1). NFMYSKTVKRLSKLREYQQYHPSLTCVMEGKDIEDWSCCPTPWTSFQSSCYFISTGMQSWTKSQKNCSVMGADLVVINTREEQDFIIQNLKRNSSYFLGLSDPGGRRHWQWVDQTPYNENVTFWHSGEPNNLDERCAIINFRSSEEWGWNDIHCHVPQKSICKMKKIYI | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 170482 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |