missing translation for 'onlineSavingsMsg'
Learn More

DLL1 (delta-like 1) Monoclonal Antibody (HMD1-5), Functional Grade, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

2985.00€ - 14910.00€

Spécification

Antigène DLL1 (delta-like 1)
Clone HMD1-5
Concentration 1 mg/mL
Applications Functional Assay
Classification Monoclonal
Consulter d'autres spécifications

Produits 3
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
17131302
View Documents
Affymetrix eBioscience
16-5767-38
5 mg Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
N/A View alternative products
Save Up to 
»
17111302
View Documents
Affymetrix eBioscience
16-5767-025
25 mg Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
N/A View alternative products
Save Up to 
»
17121302
View Documents
Affymetrix eBioscience
16-5767-050
50 mg Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
N/A View alternative products
Save Up to 
»
Description

Description

The HMD1-5 MAb reacts with mouse Delta-like 1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Delta-like 1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Delta-like 1 is expressed by thymic stromal cells, as well as on macrophages, dendritic cells, and stromal cells in the spleen. This protein is also expressed in nonhematopoietic tissues such as the lung and brain. Delta-like 1 has been linked to B cell development and differentiation, especially marginal zone and plasma cells, regulation of splenic dendritic cells, and leukemogenesis. This MAb has been reported to have neutralizing activity.

DLL1 (delta-like 1) Monoclonal antibody specifically detects DLL1 (delta-like 1) in Mouse samples. It is validated for Functional Assay
Specifications

Specifications

DLL1 (delta-like 1)
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
C-Delta-1; Delta; delta like canonical Notch ligand 1; delta like-1; Delta1; delta-like 1; delta-like 1 (Drosophila); delta-like protein 1; DL1; Dll1; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1; H-Delta-1; OTTHUMP00000017690; UNQ146/PRO172
IgG
Affinity chromatography
Antibody
HMD1-5
Functional Assay
Functional Grade
Armenian Hamster
Mouse
Q61483
13388
Primary
4° C
DLL1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.