missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase Kappa Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-57385
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DNA Polymerase Kappa Polyclonal specifically detects DNA Polymerase Kappa in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifikationer
| DNA Polymerase Kappa | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| DINB protein, DINPPOLQDINB1EC 2.7.7.7, DNA polymerase kappa, polymerase (DNA directed) kappa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51426 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| POLK | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MDSTKEKCDSYKDDLLLRMGLNDNKAGMEGLDKEKINKIIMEATKGSRFYGNELKKEKQVNQRIENMMQQKAQITSQQL | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering