missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DO-11.10 Clonotypic TCR Mouse anti-Mouse, BUV805, Clone: KJ1-26, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 750783
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DO-11.10 Clonotypic TCR Monoclonal antibody specifically detects DO-11.10 Clonotypic TCR in Mouse samples. It is validated for Flow Cytometry
Spécification
| DO-11.10 Clonotypic TCR | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| DO-11.10 clonotypic TCR | |
| DO-11.10 T hybridoma cells | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| KJ1-26 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu