Learn More
Novus Biologicals™ Dopamine beta-Hydroxylase Antibody (DBH/7222) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-26920B
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Dopamine beta-Hydroxylase Monoclonal antibody specifically detects Dopamine beta-Hydroxylase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
| Dopamine beta-Hydroxylase | |
| Monoclonal | |
| Biotin | |
| DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1 | |
| Recombinant fragment (around aa200-400) of human Dopamine beta-Hydroxylase protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Neuroscience | |
| 1621 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| DBH/7222 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |