missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPEP1 Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA543849
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a kidney membrane enzyme involved in the metabolism of glutathione and other similar proteins by dipeptide hydrolysis. The encoded protein is known to regulate leukotriene activity by catalyzing the conversion of leukotriene D4 to leukotriene E4. This protein uses zinc as a cofactor and acts as a disulfide-linked homodimer. Two transcript variants encoding the same protein have been found for this gene.
Spécification
| DPEP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P16444 | |
| DPEP1 | |
| Recombinant DPEP1 (Asp17-Leu411) expressed in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| DPEP1 | |
| AI327012; Dehydropeptidase-I; dipeptidase 1; dipeptidase 1 (renal); Dpep1; hRDP; MBD; Mbd1; MBD-1; MDP; membrane bound dipeptidase type II; membrane-bound dipeptidase 1; Microsomal dipeptidase; RDP; Renal dipeptidase; testicular tissue protein Li 57 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu