Learn More
dsRNA Antibody (1D3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-20110-0.2mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Spécification
| dsRNA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 1D3 | |
| Unconjugated | |
| Double-stranded RNA | |
| RDV-RNA-methylated bovine serum albumin complex. | |
| 0.2 mg | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG1 κ |