missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17737-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.Spécification
| EDC4 | |
| Polyclonal | |
| Unconjugated | |
| EDC4 | |
| Autoantigen Ge 1, Autoantigen RCD 8, EDC4, enhancer of mRNA decapping 4, Ge 1, HEDLS, RCD 8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234699, 23644, 361399 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UJB9, Q3ZAV8, Q6P2E9 | |
| Edc4 | |
| EDC4 Fusion Protein Ag11784 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu