Learn More
EID1 Antibody, Novus Biologicals™
Code produit 18604526 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
25 μL
0.1 mL
Conditionnement:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18604526 25 μL 25µL
18183119 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18604526 Fournisseur Novus Biologicals Code fournisseur NBP24735525ul
 Article restreint
Expédition estimée: 02-12-2026
Rabbit Polyclonal Antibody
EID1 Polyclonal specifically detects EID1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Antigène EID1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formule PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gène 21 kDa pRb-associated protein, C15orf3PTD014, CREBBP/EP300 inhibitor 1, CREBBP/EP300 inhibitory protein 1MGC138884, CRI1EP300-interacting inhibitor of differentiation 1, E1A-like inhibitor of differentiation 1, EID-1MGC138883, EP300 interacting inhibitor of differentiation 1, IRO45620, NB4 apoptosis related protein, Rb- and p300-binding protein EID-1, RBP21, retinoblastoma protein-associated protein
Symboles de gène(s) EID1
Espèces hôtes Rabbit
Immunogène This antibody was developed against a recombinant protein corresponding to amino acids: MSEMAELSELYEESSDLQMDVMPGEGDLPQMEVGSGSRELSLRPSRSGAQQLEEEGPMEEEEAQPMAAPEGKRSLANGPNAGEQPGQVAGADFESED
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Disciplines de recherche Vision
Primaire ou secondaire Primary
Identification génétique (Entrez) 23741
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Type de produit Antibody
Isotype IgG
Gamme de produits Novus
Afficher plus Afficher moins
For Research Use Only