missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2 alpha/EIF2S1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB100-2249
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
eIF2 alpha/EIF2S1 Polyclonal specifically detects eIF2 alpha/EIF2S1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Spécification
| eIF2 alpha/EIF2S1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| eIF2-alpha. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P05198 | |
| EIF2S1 | |
| The immunogen recognized by this antibody maps to a region between residue 275 and the C-terminus (residue 315) of human Eukarytic Translation Initiation Factor 1, subunit 1 alpha, 35kDa using the numbering given in entry NP_004085.1 (GeneID 1965). | |
| Affinity Purified | |
| RUO | |
| 1965 | |
| Human, Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu