missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastase Antibody (CELA3B/1811), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-54489MFV500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Elastase Monoclonal antibody specifically detects Elastase in Human samples. It is validated for Flow Cytometry
Spécification
| Elastase | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| CELA3B/1811 | |
| Flow Cytometry | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Recombinant human Elastase protein fragment (around aa 82-238) (exact sequence is proprietary) (Uniprot: P08861) | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23436 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu