missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Elastin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA553727
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: VKPGKVPGVG LPGVYPGGVL PGARFPGVGV LPGVPTGAGV KPKAPGVGGA FAGIPGVGPF GGPQPGVPLG YPIKAPKLPG GYGLPYTTGK LPYGYGPGGV AGAAGKAGYP TGTGVGPQAA AAAAAKAAAK FGAGA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification
| Elastin | |
| Polyclonal | |
| Unconjugated | |
| ELN | |
| 5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2006 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15502 | |
| ELN | |
| Recombinant protein corresponding to Human Elastin. Recombinant protein control fragment (Product #RP-91206). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu