Learn More
ELF3 Monoclonal Antibody (OTI4G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA809988
Description
ESE-1 (Epithelium-Specific Ets) is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family, which has features distinct from those of any other ets-related factor. ESE-1 binds with high affinity to and transactivates the ets binding site in the promoter of the keratinocyte terminal differentiation marker gene, SPRR2A. ESE-1 is a critical regulator of epithelial cell differentiation.Spécification
| ELF3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78545 | |
| ELF3 | |
| Human recombit protein fragment corresponding to amino acids 175-286 of human ELF3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G5 | |
| Unconjugated | |
| ELF3 | |
| EPR-1, ERT, ESE-1, ESX | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1999 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.