Learn More
ELTD1/ETL Antibody (CL4164) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP3-44095
Code nomenclature Nacres: NA.46
Code produit. 30289432
  • 638.00€ / 100µL
Description
ELTD1/ETL Monoclonal antibody specifically detects ELTD1/ETL in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Spécification
ELTD1/ETL
Monoclonal
LYOPH
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003
This antibody was generated using a recombinant protein sequence of Q9HBW9, with the exact immunogen sequence remaining proprietary.
100 μL
GPCR, Neuroscience
64123
Human
Purified
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
CL4164
Unconjugated
Lyophilized in 5% Trehalose.
Mouse
Protein A purified
RUO
Primary
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
IgG1