Description
EME1 Monoclonal specifically detects EME1 in Human samples. It is validated for Western Blot.
Spécification
Spécification
| Antigène | EME1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MTA31 7h2/1 |
| Conjugué | DyLight 550 |
| Dilution | Western Blot |
| Formule | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gène | crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A |
| Symboles de gène(s) | EME1 |
| Espèces hôtes | Mouse |
| Afficher plus |