Learn More
Endoglin/CD105 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30730-1-AP-150UL
Nacres classification code: NA.46
 View more versions of this product
Product Code. 18016695
  • 410.00€ / 150µL
This item is not returnable. View return policy
Description
Description
The Endoglin/CD105 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Endoglin/CD105. This antibody recognizes human antigen. The Endoglin/CD105 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Endoglin/CD105
550 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
CD105, END, endoglin, ENG, FLJ41744, HHT1, ORW, ORW1
150 μL
2022
Store at -20°C. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS