Learn More
Endoglin/CD105 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30730-1-AP-150UL
Description
The Endoglin/CD105 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Endoglin/CD105. This antibody recognizes human antigen. The Endoglin/CD105 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Endoglin/CD105 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| CD105, END, endoglin, ENG, FLJ41744, HHT1, ORW, ORW1 | |
| 150 μL | |
| 2022 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.