missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endothelin 1 Monoclonal Antibody (ENDO20-2101.70)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA110818
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-10818 detects Endothelin from human samples. MA1-10818 has been successfully used in ELISA, radioimmune assay, and immunohistochemistry applications. The MA1-10818 immunogen is Human endothelin 1.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
Spécification
| Endothelin 1 | |
| Monoclonal | |
| 1.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P05305 | |
| EDN1 | |
| Human endothelin 1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
| ENDO20-2101.70 | |
| Unconjugated | |
| EDN1 | |
| ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME | |
| Mouse | |
| Protein G | |
| RUO | |
| 1906 | |
| -20°C or -80°C if preferred | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu