missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENT1 Polyclonal Antibody

ENT1 Polyclonal Antibody
Marque: Invitrogen PA589074
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ENT1 Polyclonal Antibody for Western Blot, IHC (P)
Spécification
| ENT1 | |
| Polyclonal | |
| Unconjugated | |
| SLC29A1 | |
| 1200014D21Rik; AA407560; ENT1; Equilibrative NBMPR-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside (NBMPR)-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter; Equilibrative nucleoside transporter 1; equilibrative nucleoside transporter 1 variant delta 11; HENT1; NBMPR-sensitive equilibrative nucleoside transporter; nucleoside transporter, es-type; rENT1; SLC29A1; solute carrier family 29 (equilibrative nucleoside transporter), member 1; solute carrier family 29 (nucleoside transporters), member 1; solute carrier family 29 member 1; solute carrier family 29 member 1 (Augustine blood group); solute carrier family 29, member 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2030 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Western Blot | |
| 1.31 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99808 | |
| SLC29A1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SLC29A1 (NP_0010716451). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu