Learn More
ENT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque:  Novus Biologicals NBP1-85253
Code nomenclature Nacres: NA.46
Code produit. 18701584
  • 572.00€ / 0.10mL
Description
ENT2 Polyclonal specifically detects ENT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Polyclonal
Delayed-early response protein 12, DER12Solute carrier family 29 member 2, ENT2, Equilibrative NBMPR-insensitive nucleoside transporter, Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleosidetransporter, equilibrative nucleoside transporter 2, HNP3636 kDa nucleolar protein HNP36, Hydrophobic nucleolar protein, 36 kDa, hydrophobic nucleolar protein, 36kD, Nucleoside transporter, ei-type, solute carrier family 29 (nucleoside transporters), member 2
0.1 mL
IgG
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
This antibody was developed against Recombinant Protein corresponding to amino acids:KFARYYLANKSSQAQAQELETKAELLQSDENGIPSSPQKVALTLDLDLEKEPESEPDEPQKPGKPSVFTVFQK
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
For Research Use Only